Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPOCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281987.1 | NP_001268916.1 | 9 | 2208 | ||
NM_001281988.1 | NP_001268917.1 | 8 | 786 | |||
NM_144569.5 | NP_653170.3 | 9 | 2208 | |||
XM_011542413.2 | XP_011540715.1 | 9 | 777 | |||
XM_011542414.2 | XP_011540716.1 | 8 | 972 | |||
XM_017002779.1 | XP_016858268.1 | 8 | 2117 | |||
XM_017002780.1 | XP_016858269.1 | 7 | 2075 | |||
XM_017002781.1 | XP_016858270.1 | 8 | 2117 | |||
XM_017002782.1 | XP_016858271.1 | 8 | 1346 | |||
GenBank mRNA | AK091016.1 | - | 8 | 757 | ||
AK095829.1 | - | 2 | 384 | |||
AK096192.1 | - | 1 | 358 | |||
AK097227.1 | - | 6 | 612 | |||
AK131447.1 | - | 9 | 2137 | |||
BC007910.2 | - | 2 | 274 | |||
BC029069.1 | - | 2 | 224 | |||
BC114430.1 | - | 9 | 2134 | 40038407 |