Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017989.2 | NP_001017989.2 | 1 | 155 | ||
NM_025136.3 | NP_079412.1 | 1 | 155 | |||
GenBank mRNA | AK025840.1 | - | 1 | 110 | ||
AK098798.1 | - | 1 | 133 | |||
BC005059.1 | - | 1 | 76 | 2957964 | ||
BC047316.1 | - | 1 | 76 | 5206005 | ||
BC064146.1 | - | 1 | 67 | 30378779 | ||
DB034102.1 | - | 1 | 155 |