Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016466.5 | NP_057550.3 | 4 | 623 | ||
GenBank mRNA | AF151034.1 | - | 4 | 604 | ||
BC031303.2 | - | 4 | 615 | 5284947 |