Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANAPC11 SIRNA Assays ›
Gene Symbol:
ANAPC11
Gene Name:
anaphase promoting complex subunit 11
Gene Aliases:
APC11, Apc11p, HSPC214
Chromosome Location:
Chr.17: 81890790 - 81900533 on Build GRCh38
UniGene ID:
Hs.534456
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
24377
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANAPC11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002244.2
NP_001002244.1
2
250
NM_001002245.2
NP_001002245.1
2
220
NM_001002246.2
NP_001002246.1
3
283
NM_001002247.2
NP_001002247.1
2
228
NM_001002248.2
NP_001002248.1
3
291
NM_001002249.2
NP_001002249.1
3
342
NM_001289414.1
NP_001276343.1
3
224
NM_001289415.1
NP_001276344.1
2
250
NM_001289416.1
NP_001276345.1
2
279
NM_001289417.1
NP_001276346.1
4
375
NM_001289418.1
NP_001276347.1
2
220
NM_001289419.1
NP_001276348.1
3
291
NM_001289420.1
NP_001276349.1
3
342
NM_016476.11
NP_057560.8
3
313
GenBank mRNA
AA356010.1
-
3
225
AF151048.1
-
3
258
AF247565.1
-
2
158
AF247789.1
-
3
236
AK307214.1
-
3
283
AK308091.1
-
1
134
AW965775.1
-
3
224
BC000607.1
-
2
226
3346039
BC066308.1
-
3
327
BC095454.1
-
3
242
BC104641.1
-
2
176
BC171892.1
-
3
225
BC171898.1
-
2
192
BC171899.1
-
3
233
BC171900.1
-
2
162
BG707824.1
-
3
274
BM762129.1
-
3
259
BU554167.1
-
3
293
BU556841.1
-
3
257
BY796995.2
-
2
143
CA335305.1
-
4
338
CB106395.1
-
2
218
DB501853.1
-
2
210
DB573049.1
-
1
56
HY067392.1
-
2
221
HY142453.1
-
3
270
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
mitotic nuclear division
protein ubiquitination
anaphase-promoting complex-dependent catabolic process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of mitotic metaphase/anaphase transition
cell division
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
protein K11-linked ubiquitination
ubiquitin-protein transferase activity
zinc ion binding
ubiquitin-ubiquitin ligase activity
ubiquitin protein ligase activity
cullin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0