Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016481.4 | NP_057565.3 | 1, 2 | 161 | ||
XM_005252047.3 | XP_005252104.1 | 1, 2 | 118 | |||
XM_005252048.4 | XP_005252105.1 | 1, 2 | 118 | |||
XM_011518776.2 | XP_011517078.1 | 1, 2 | 118 | |||
GenBank mRNA | AF151053.1 | - | 1, 2 | 108 | ||
AK000213.1 | - | 1, 2 | 161 | |||
AK302701.1 | - | 1, 2 | 161 | |||
AK309270.1 | - | 1, 2 | 120 | |||
AY189280.1 | - | 1, 2 | 108 | |||
BC002863.2 | - | 1, 2 | 141 | 3944148 |