Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016486.3 | NP_057570.2 | 3 | 658 | ||
GenBank mRNA | AF151063.1 | - | 3 | 637 | ||
AK024682.1 | - | 3 | 652 | |||
BC013608.1 | - | 3 | 640 | 3864518 | ||
BC034147.1 | - | 3 | 638 | 4655713 | ||
BC039718.2 | - | 2 | 561 | 5784175 | ||
BC046130.1 | - | 3 | 621 | 5787422 | ||
BC054041.1 | - | 1 | 372 | |||
BC064539.1 | - | 3 | 641 | 5725426 | ||
BC073836.1 | - | 2 | 558 | 5548608 | ||
BC121117.2 | - | 3 | 600 | 40120483 | ||
BC133692.1 | - | 3 | 599 | 40120480 |