Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291963.1 | NP_001278892.1 | 3 | 557 | ||
NM_004394.2 | NP_004385.1 | 4 | 600 | |||
GenBank mRNA | AK298670.1 | - | 3 | 510 | ||
BC002726.2 | - | 4 | 557 | 3631886 | ||
BI823002.1 | - | 4 | 430 | |||
BP265421.1 | - | 4 | 561 | |||
BT006925.1 | - | 3 | 266 | |||
X76105.1 | - | 4 | 552 |