Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKX2-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079668.2 | NP_001073136.1 | 2 | 215 | ||
NM_003317.3 | NP_003308.1 | 1 | 370 | |||
GenBank mRNA | BC006221.2 | - | 2 | 149 | 3941576 | |
BE796718.1 | - | 2 | 130 | |||
BT009773.1 | - | 2 | 119 | |||
D50740.1 | - | 3 | 273 | |||
D50741.1 | - | 1 | 90 | |||
U19756.1 | - | 1 | 29 | |||
U33749.1 | - | 1 | 370 | |||
U43203.1 | - | 2 | 201 | |||
X82850.1 | - | 1 | 43 |