siRNA Details
Gene Symbol: SP110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185015.1 | NP_001171944.1 | 12 | 1539 | ||
NM_004509.3 | NP_004500.3 | 11 | 1503 | |||
NM_004510.3 | NP_004501.3 | 11 | 1503 | |||
NM_080424.2 | NP_536349.2 | 11 | 1503 | |||
XM_005246525.3 | XP_005246582.1 | 12 | 1539 | |||
XM_006712487.3 | XP_006712550.1 | 12 | 1539 | |||
XM_006712489.3 | XP_006712552.1 | 12 | 1539 | |||
XM_011511088.2 | XP_011509390.1 | 12 | 1539 | |||
XM_011511089.2 | XP_011509391.1 | 11 | 1394 | |||
XM_011511090.2 | XP_011509392.1 | 11 | 1389 | |||
XM_011511091.2 | XP_011509393.1 | 12 | 1539 | |||
XM_011511092.2 | XP_011509394.1 | 10 | 1232 | |||
XM_017003968.1 | XP_016859457.1 | 13 | 1614 | |||
XM_017003969.1 | XP_016859458.1 | 11 | 1389 | |||
XM_017003970.1 | XP_016859459.1 | 10 | 1244 | |||
GenBank mRNA | AF280094.1 | - | 11 | 1340 | ||
AF280095.1 | - | 11 | 1340 | |||
AK026488.1 | - | 11 | 1342 | |||
AK301097.1 | - | 12 | 1539 | |||
AL832300.1 | - | 10 | 1277 | |||
BC019059.2 | - | 11 | 1358 | 5088115 | ||
L22343.1 | - | 8 | 824 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
