Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137604.2 | NP_001131076.1 | 13 | 2742 | ||
NM_001282772.1 | NP_001269701.1 | 15 | 2560 | |||
NM_001282774.1 | NP_001269703.1 | 12 | 2361 | |||
NM_001282776.1 | NP_001269705.1 | 11 | 2462 | |||
NM_001282777.1 | NP_001269706.1 | 15 | 3002 | |||
NM_001282779.1 | NP_001269708.1 | 14 | 2839 | |||
NM_019014.5 | NP_061887.2 | 14 | 2910 | |||
GenBank mRNA | AK001678.1 | - | 14 | 2382 | ||
AK022533.1 | - | 13 | 2192 | |||
AK022890.1 | - | 7 | 1073 | |||
AK025574.1 | - | 3 | 418 | |||
AK128044.1 | - | 14 | 2910 | |||
AK294027.1 | - | 14 | 2311 | |||
AK301178.1 | - | 11 | 1934 | |||
AK302814.1 | - | 15 | 2560 | |||
BC004882.2 | - | 5 | 672 | |||
BC073857.1 | - | 5 | 606 | |||
BC096748.1 | - | 5 | 707 | |||
BC110833.1 | - | 12 | 2050 | 6053210 | ||
BC150139.1 | - | 6 | 1116 | |||
BX647360.1 | - | 15 | 2728 | |||
BX647683.1 | - | 14 | 2289 |