Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145418.1 | NP_001138890.1 | 23 | 7328 | ||
XM_005261405.2 | XP_005261462.1 | 22 | 7304 | |||
XM_006724171.3 | XP_006724234.1 | 22 | 16106 | |||
XM_011530018.2 | XP_011528320.1 | 23 | 7507 | |||
XM_017028673.1 | XP_016884162.1 | 23 | 7301 | |||
GenBank mRNA | AB665284.1 | - | 23 | 7276 | ||
AL080174.1 | - | 2 | 1459 | |||
AL137328.1 | - | 1 | 96 | |||
BC016465.1 | - | 15 | 3872 | 4154050 |