Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABI2 SIRNA Assays ›
Gene Symbol:
ABI2
Gene Name:
abl-interactor 2
Gene Aliases:
ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
Chromosome Location:
Chr.2: 203327874 - 203432173 on Build GRCh38
UniGene ID:
Hs.471156
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
244115
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABI2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282925.1
NP_001269854.1
5
787
NM_001282926.1
NP_001269855.1
4
769
NM_001282927.1
NP_001269856.1
4
445
NM_001282932.1
NP_001269861.1
5
966
NM_005759.5
NP_005750.4
4
769
XM_005246217.2
XP_005246274.1
5
1152
XM_005246218.2
XP_005246275.1
4
1134
XM_005246221.2
XP_005246278.1
4
1134
XM_005246225.2
XP_005246282.1
4
1134
XM_005246226.2
XP_005246283.1
4
1134
XM_006712174.2
XP_006712237.1
5
1152
XM_006712177.2
XP_006712240.1
6
1228
XM_006712179.2
XP_006712242.1
5
1152
XM_006712180.2
XP_006712243.1
5
1152
XM_006712181.2
XP_006712244.1
5
1152
XM_006712182.2
XP_006712245.1
5
1152
XM_006712185.1
XP_006712248.1
5
1210
XM_006712186.2
XP_006712249.1
4
1134
XM_011510472.2
XP_011508774.1
5
1152
XM_011510473.2
XP_011508775.1
4
451
XM_017003118.1
XP_016858607.1
5
1152
XM_017003119.1
XP_016858608.1
5
1152
XM_017003120.1
XP_016858609.1
5
1152
XM_017003121.1
XP_016858610.1
5
1152
XM_017003122.1
XP_016858611.1
5
1152
XM_017003123.1
XP_016858612.1
4
1134
XM_017003124.1
XP_016858613.1
4
1134
XM_017003125.1
XP_016858614.1
4
1134
XM_017003126.1
XP_016858615.1
4
1134
XM_017003127.1
XP_016858616.1
4
1134
XM_017003128.1
XP_016858617.1
6
1228
XM_017003129.1
XP_016858618.1
5
1152
XM_017003130.1
XP_016858619.1
5
1152
XM_017003131.1
XP_016858620.1
4
1134
XM_017003132.1
XP_016858621.1
5
1152
XM_017003133.1
XP_016858622.1
6
1228
XM_017003134.1
XP_016858623.1
4
1134
XM_017003135.1
XP_016858624.1
4
1134
XM_017003136.1
XP_016858625.1
4
1134
XM_017003137.1
XP_016858626.1
6
1228
XM_017003138.1
XP_016858627.1
5
1152
XM_017003139.1
XP_016858628.1
5
1210
XM_017003140.1
XP_016858629.1
5
1210
XM_017003141.1
XP_016858630.1
4
1134
XM_017003142.1
XP_016858631.1
4
1134
XM_017003143.1
XP_016858632.1
6
1228
XM_017003144.1
XP_016858633.1
5
1152
XM_017003145.1
XP_016858634.1
4
1134
XM_017003146.1
XP_016858635.1
4
1134
GenBank mRNA
AF260261.1
-
5
511
AK297537.1
-
2
58
AK299092.1
-
3
71
AK299672.1
-
3
77
AK299824.1
-
4
613
AK302853.1
-
5
892
AK316000.1
-
7
726
BC001439.2
-
4
518
3138341
BT009920.1
-
4
493
U23435.1
-
4
406
U31089.1
-
3
238
X95632.1
-
4
593
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
movement of cell or subcellular component
cytoskeleton organization
actin polymerization or depolymerization
viral process
cell migration
Rac protein signal transduction
peptidyl-tyrosine phosphorylation
positive regulation of Arp2/3 complex-mediated actin nucleation
DNA binding
protein binding
cytoskeletal adaptor activity
SH3 domain binding
kinase binding
ubiquitin protein ligase binding
protein complex binding
Rac GTPase binding
proline-rich region binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0