Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122870.2 | NP_001116342.1 | 9 | 1062 | ||
NM_144641.3 | NP_653242.3 | 10 | 1428 | |||
XM_005264879.2 | XP_005264936.1 | 10 | 1521 | |||
XM_005264880.3 | XP_005264937.1 | 9 | 1241 | |||
XM_005264881.1 | XP_005264938.1 | 10 | 1317 | |||
GenBank mRNA | AK056894.1 | - | 9 | 1664 | ||
AK096681.1 | - | 8 | 1356 | |||
AK129647.1 | - | 4 | 693 | |||
AL832484.1 | - | 8 | 1336 | |||
BC009644.1 | - | 9 | 1023 | 3905720 |