Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIEZO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022068.3 | NP_071351.2 | 19, 20 | 3049 | ||
XM_011525723.2 | XP_011524025.1 | 22, 23 | 3979 | |||
XM_011525724.2 | XP_011524026.1 | 21, 22 | 3922 | |||
XM_011525725.2 | XP_011524027.1 | 20, 21 | 3889 | |||
XM_011525726.2 | XP_011524028.1 | 22, 23 | 3979 | |||
XM_017025918.1 | XP_016881407.1 | 21, 22 | 3940 | |||
GenBank mRNA | AK127627.1 | - | 2, 3 | 339 | ||
JN790819.1 | - | 19, 20 | 2883 |