Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCP11L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145541.1 | NP_001139013.1 | 7 | 1117 | ||
NM_018393.3 | NP_060863.3 | 7 | 1301 | |||
XM_011520204.2 | XP_011518506.1 | 7 | 1132 | |||
XM_011520205.2 | XP_011518507.1 | 7 | 1300 | |||
XM_011520206.2 | XP_011518508.1 | 7 | 1318 | |||
XM_011520207.2 | XP_011518509.1 | 7 | 1314 | |||
XM_017017989.1 | XP_016873478.1 | 7 | 1315 | |||
XM_017017990.1 | XP_016873479.1 | 7 | 1285 | |||
XM_017017991.1 | XP_016873480.1 | 5 | 678 | |||
XM_017017992.1 | XP_016873481.1 | 5 | 736 | |||
GenBank mRNA | AK002198.1 | - | 7 | 1099 | ||
AK021448.1 | - | 2 | 263 | |||
AK090646.1 | - | 7 | 1284 | |||
BC041696.1 | - | 7 | 1120 | 5756130 |