Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033402.4 | NP_208325.3 | 12 | 2136 | ||
XM_005251318.3 | XP_005251375.1 | 11 | 1925 | |||
XM_005251319.4 | XP_005251376.1 | 10 | 1619 | |||
XM_005251320.3 | XP_005251377.1 | 9 | 1411 | |||
XM_005251322.3 | XP_005251379.1 | 8 | 1225 | |||
XM_017013920.1 | XP_016869409.1 | 13 | 2084 | |||
XM_017013921.1 | XP_016869410.1 | 13 | 2084 | |||
XM_017013922.1 | XP_016869411.1 | 13 | 2084 | |||
XM_017013923.1 | XP_016869412.1 | 12 | 1911 | |||
XM_017013924.1 | XP_016869413.1 | 13 | 2084 | |||
XM_017013925.1 | XP_016869414.1 | 10 | 1630 | |||
XM_017013926.1 | XP_016869415.1 | 10 | 1613 | |||
XM_017013927.1 | XP_016869416.1 | 11 | 1641 | |||
XM_017013928.1 | XP_016869417.1 | 8 | 1093 | |||
GenBank mRNA | AB051551.1 | - | 11 | 2757 | ||
AK056185.1 | - | 11 | 2099 | |||
AK057990.1 | - | 3 | 391 | |||
AK300234.1 | - | 9 | 1368 | |||
AK300874.1 | - | 11 | 1780 | |||
AK302648.1 | - | 12 | 1986 | |||
AK307443.1 | - | 9 | 1392 | |||
AK307636.1 | - | 11 | 1770 | |||
BC030701.2 | - | 12 | 2009 | 4837403 |