Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094396.1 | - | 1 | 380 |
Gene Symbol: TRIM74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012197.1 | XP_016867686.1 | 8 | 1409 |