Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039550.1 | NP_001034639.1 | 2 | 282 | ||
NM_006736.5 | NP_006727.2 | 2 | 282 | |||
GenBank mRNA | AK289626.1 | - | 2 | 105 | ||
AK292761.1 | - | 2 | 110 | |||
AK293888.1 | - | 2 | 234 | |||
AK312723.1 | - | 2 | 289 | |||
AL547521.3 | - | 2 | 328 | |||
BC011609.2 | - | 2 | 224 | 3619734 | ||
BC047056.1 | - | 1 | 85 | 5185009 | ||
BT007088.1 | - | 1 | 45 | |||
BX420085.2 | - | 284 | ||||
S37374.1 | - | 1 | 70 | |||
S37375.1 | - | 1 | 70 |