Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CABIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199281.1 | NP_001186210.1 | 34 | 6242 | ||
NM_001201429.1 | NP_001188358.1 | 33 | 5786 | |||
NM_012295.3 | NP_036427.1 | 34 | 5936 | |||
XM_005261415.2 | XP_005261472.1 | 33 | 5988 | |||
XM_005261416.2 | XP_005261473.1 | 33 | 5973 | |||
XM_005261417.2 | XP_005261474.1 | 33 | 5898 | |||
XM_005261419.2 | XP_005261476.1 | 33 | 6029 | |||
XM_006724179.1 | XP_006724242.1 | 33 | 5988 | |||
XM_006724180.1 | XP_006724243.1 | 33 | 5811 | |||
XM_006724181.1 | XP_006724244.1 | 33 | 5883 | |||
XM_011530026.1 | XP_011528328.1 | 32 | 5793 | |||
XM_011530029.2 | XP_011528331.1 | 33 | 5988 | |||
XM_017028680.1 | XP_016884169.1 | 33 | 5973 | |||
XM_017028681.1 | XP_016884170.1 | 32 | 5838 | |||
XM_017028682.1 | XP_016884171.1 | 33 | 5909 | |||
XM_017028683.1 | XP_016884172.1 | 32 | 5646 | |||
XM_017028684.1 | XP_016884173.1 | 18 | 3526 | |||
GenBank mRNA | AB002328.2 | - | 34 | 6159 | ||
AF072441.1 | - | 34 | 5937 | |||
AL390166.1 | - | 6 | 1133 | |||
BC021929.1 | - | 9 | 1716 | |||
BC054497.1 | - | 33 | 5666 | 4521929 |