Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CABIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199281.1 | NP_001186210.1 | 37 | 7273 | ||
NM_001201429.1 | NP_001188358.1 | 36 | 6817 | |||
NM_012295.3 | NP_036427.1 | 37 | 6967 | |||
XM_005261415.2 | XP_005261472.1 | 36 | 7019 | |||
XM_005261416.2 | XP_005261473.1 | 36 | 7004 | |||
XM_005261417.2 | XP_005261474.1 | 36 | 6929 | |||
XM_005261419.2 | XP_005261476.1 | 36 | 7060 | |||
XM_006724179.1 | XP_006724242.1 | 36 | 6932 | |||
XM_006724180.1 | XP_006724243.1 | 36 | 6842 | |||
XM_006724181.1 | XP_006724244.1 | 36 | 6827 | |||
XM_011530026.1 | XP_011528328.1 | 35 | 6824 | |||
XM_017028680.1 | XP_016884169.1 | 36 | 6917 | |||
XM_017028681.1 | XP_016884170.1 | 35 | 6869 | |||
XM_017028682.1 | XP_016884171.1 | 36 | 6940 | |||
XM_017028683.1 | XP_016884172.1 | 35 | 6677 | |||
XM_017028684.1 | XP_016884173.1 | 21 | 4557 | |||
GenBank mRNA | AB002328.2 | - | 37 | 7190 | ||
AF072441.1 | - | 37 | 6968 | |||
AK127950.1 | - | 1 | 2461 | |||
AL390166.1 | - | 8 | 1999 | |||
BC021929.1 | - | 12 | 2747 | |||
BC054497.1 | - | 36 | 6697 | 4521929 |