Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286420.1 | NP_001273349.1 | 3 | 459 | ||
NM_016454.3 | NP_057538.1 | 3 | 459 | |||
GenBank mRNA | AF151018.1 | - | 3 | 412 | ||
AK075227.1 | - | 3 | 438 | |||
AK290524.1 | - | 3 | 443 | |||
AY336092.1 | - | 3 | 399 | |||
BC002583.2 | - | 3 | 424 | 3161718 | ||
KU178640.1 | - | 3 | 341 | |||
KU178641.1 | - | 3 | 341 |