Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCOA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122842.2 | NP_001116314.1 | 16 | 4224 | ||
NM_001199619.1 | NP_001186548.1 | 17 | 4240 | |||
NM_001199620.1 | NP_001186549.1 | 18 | 4358 | |||
NM_001199621.1 | NP_001186550.1 | 15 | 4003 | |||
NM_001199622.1 | NP_001186551.1 | 6 | 1852 | |||
NM_181782.4 | NP_861447.3 | 16 | 4257 | |||
XM_005266822.4 | XP_005266879.1 | 16 | 4287 | |||
XM_006715340.3 | XP_006715403.1 | 16 | 4214 | |||
XM_011535455.2 | XP_011533757.1 | 16 | 4162 | |||
XM_017010269.1 | XP_016865758.1 | 16 | 3973 | |||
XM_017010270.1 | XP_016865759.1 | 17 | 4186 | |||
XM_017010271.1 | XP_016865760.1 | 16 | 4184 | |||
XM_017010272.1 | XP_016865761.1 | 16 | 4037 | |||
XM_017010273.1 | XP_016865762.1 | 16 | 4254 | |||
XM_017010274.1 | XP_016865763.1 | 16 | 4343 | |||
GenBank mRNA | AF493978.1 | - | 17 | 4239 | ||
AJ420542.1 | - | 8 | 2189 | |||
AK094706.1 | - | 6 | 1745 | |||
AL832621.2 | - | 16 | 4017 | |||
AL832628.2 | - | 16 | 3979 | |||
AL834442.1 | - | 14 | 3644 | |||
BC033612.1 | - | 9 | 3084 | |||
BX537385.1 | - | 16 | 4052 | |||
KC238672.1 | - | 6 | 1829 |