Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164114.1 | NP_001157586.1 | 2 | 416 | ||
NM_001164115.1 | NP_001157587.1 | 2 | 416 | |||
NM_001164116.1 | NP_001157588.1 | 3 | 440 | |||
NM_020356.3 | NP_065089.2 | 2 | 416 | |||
XM_006723831.3 | XP_006723894.1 | 3 | 1002 | |||
GenBank mRNA | AJ276678.1 | - | 3 | 440 | ||
AK027760.1 | - | 2 | 413 | |||
AK295613.1 | - | 2 | 281 | |||
AK308057.1 | - | 2 | 417 | |||
AK314653.1 | - | 2 | 349 | |||
BC027951.1 | - | 2 | 366 | 5205865 | ||
BX280195.1 | - | 2 | 366 | |||
DA584687.1 | - | 2 | 417 |