Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP4M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004722.3 | NP_004713.2 | 6, 7 | 746 | ||
XM_005250689.4 | XP_005250746.1 | 7, 8 | 650 | |||
XM_005250690.4 | XP_005250747.1 | 4, 5 | 542 | |||
XM_006716175.3 | XP_006716238.1 | 6, 7 | 728 | |||
XM_011516685.2 | XP_011514987.1 | 6, 7 | 728 | |||
XM_017012790.1 | XP_016868279.1 | 5, 6 | 724 | |||
XM_017012791.1 | XP_016868280.1 | 4, 5 | 332 | |||
GenBank mRNA | AF020796.1 | - | 6, 7 | 697 | ||
AF155158.1 | - | 6, 7 | 582 | |||
AK055686.1 | - | 5, 6 | 723 | |||
AK293590.1 | - | 5, 6 | 515 | |||
AK296651.1 | - | 5, 6 | 502 | |||
AK304489.1 | - | 5, 6 | 437 | |||
AK315782.1 | - | 6, 7 | 563 | |||
BC018705.1 | - | 6, 7 | 589 | 5013882 | ||
BX640759.1 | - | 5, 6 | 495 | |||
Y08387.1 | - | 6, 7 | 592 |