Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP4M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004722.3 | NP_004713.2 | 15 | 1433 | ||
XM_005250689.4 | XP_005250746.1 | 16 | 1337 | |||
XM_005250690.4 | XP_005250747.1 | 13 | 1229 | |||
XM_011516685.2 | XP_011514987.1 | 15 | 1415 | |||
XM_017012791.1 | XP_016868280.1 | 12 | 952 | |||
GenBank mRNA | AF020796.1 | - | 15 | 1384 | ||
AF155158.1 | - | 15 | 1269 | |||
AK055686.1 | - | 14 | 1411 | |||
AK296651.1 | - | 14 | 1189 | |||
AK315782.1 | - | 15 | 1175 | |||
BC018705.1 | - | 15 | 1276 | 5013882 | ||
BX640759.1 | - | 13 | 1115 | |||
Y08387.1 | - | 15 | 1279 |