Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22orf29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024627.5 | NP_078903.3 | 2 | 252 | ||
GenBank mRNA | AK291262.1 | - | 2 | 220 | ||
AL832509.1 | - | 2 | 141 | |||
BX640785.1 | - | 1 | 341 | |||
BX640969.1 | - | 2 | 221 | |||
BX640998.1 | - | 2 | 221 | |||
DA182195.1 | - | 2 | 230 | |||
DA757523.1 | - | 2 | 220 |
Gene Symbol: GNB1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053004.2 | NP_443730.1 | 2 | 183 | ||
GenBank mRNA | AB051432.1 | - | 2 | 124 | ||
AF238328.1 | - | 2 | 183 | |||
AF301895.1 | - | 2 | 148 | |||
AY007378.1 | - | 2 | 183 | |||
BC012060.1 | - | 2 | 84 | 4548339 |
Set Membership: |
G-protein coupled receptor |