Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007533.3 | NP_001007534.1 | 1 | 12 | ||
GenBank mRNA | AK096888.1 | - | 1 | 12 | ||
DA894509.1 | - | 1 | 12 |