Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTRF1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114184.2 | NP_001107656.1 | 3 | 577 | ||
NM_001301871.1 | NP_001288800.1 | 3 | 552 | |||
NM_019041.6 | NP_061914.3 | 3 | 577 | |||
NR_126056.1 | 3 | 552 | ||||
XM_017010959.1 | XP_016866448.1 | 4 | 5184 | |||
XM_017010960.1 | XP_016866449.1 | 4 | 593 | |||
GenBank mRNA | AK027684.1 | - | 3 | 422 | ||
AK095240.1 | - | 3 | 487 | |||
AK225321.1 | - | 3 | 451 | |||
AK225970.1 | - | 3 | 451 | |||
AK290662.1 | - | 3 | 451 | |||
BC011873.2 | - | 3 | 474 | 3029407 | ||
BC014428.1 | - | 3 | 463 | 4646472 | ||
BX376520.2 | - | 3 | 445 |