Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000039.2 | NP_000030.1 | 2 | 394 | ||
NM_001318017.1 | NP_001304946.1 | 2 | 107 | |||
NM_001318018.1 | NP_001304947.1 | 2 | 99 | |||
NM_001318021.1 | NP_001304950.1 | 2 | 394 | |||
GenBank mRNA | A14829.1 | - | 1 | 47 | ||
AK292231.1 | - | 2 | 68 | |||
AL531353.3 | - | 2 | 99 | |||
AL557908.3 | - | 2 | 107 | |||
BC005380.1 | - | 2 | 66 | 3934992 | ||
BC110286.1 | - | 2 | 89 | 5164525 | ||
BU600916.1 | - | 2 | 69 | |||
CD558050.1 | - | 2 | 394 | |||
M11791.1 | - | 1 | 28 | |||
M27875.1 | - | 1 | 47 | |||
T68127.1 | - | 2 | 107 | |||
X00566.1 | - | 1 | 63 | |||
X02162.1 | - | 2 | 114 |