Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACYP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107.4 | NP_001098.1 | 2 | 145 | ||
NM_001302616.1 | NP_001289545.1 | 2 | 127 | |||
NM_001302617.1 | NP_001289546.1 | 2 | 323 | |||
NR_126393.1 | 2 | 145 | ||||
GenBank mRNA | AK312101.1 | - | 2 | 145 | ||
BC035568.1 | - | 2 | 130 | 5392248 | ||
BG614847.1 | - | 2 | 112 | |||
BP316920.1 | - | 2 | 108 | |||
CD675334.1 | - | 2 | 332 | |||
DB071115.1 | - | 2 | 145 | |||
X84194.1 | - | 1 | 109 |