Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093770.2 | NP_001087239.2 | 5 | 436 | ||
NM_001164644.1 | NP_001158116.1 | 5 | 406 | |||
NM_001164645.1 | NP_001158117.1 | 5 | 292 | |||
NM_001164646.1 | NP_001158118.1 | 5 | 300 | |||
NM_001164647.1 | NP_001158119.1 | 4 | 376 | |||
NM_005411.4 | NP_005402.3 | 5 | 447 | |||
XM_005270062.4 | XP_005270119.1 | 4 | 436 | |||
XM_006717953.2 | XP_006718016.1 | 5 | 429 | |||
GenBank mRNA | AK290703.1 | - | 5 | 438 | ||
AK298002.1 | - | 5 | 292 | |||
AK298029.1 | - | 4 | 325 | |||
AK298034.1 | - | 4 | 183 | |||
AK298092.1 | - | 3 | 178 | |||
AK316196.1 | - | 4 | 100 | |||
BC029913.2 | - | 4 | 403 | |||
BC111570.1 | - | 4 | 355 | 40069324 | ||
BC171875.1 | - | 4 | 366 | 9054529 | ||
BI820937.1 | - | 5 | 430 | |||
DA588383.1 | - | 5 | 300 | |||
HQ021433.2 | - | 4 | 376 | |||
HQ021434.1 | - | 4 | 376 | |||
HQ021435.1 | - | 4 | 376 | |||
HQ021436.1 | - | 4 | 376 | |||
HQ021437.1 | - | 5 | 447 | |||
HQ021438.1 | - | 5 | 447 | |||
HQ021439.1 | - | 5 | 447 | |||
HQ021440.1 | - | 5 | 436 | |||
HQ021441.1 | - | 5 | 436 | |||
HQ021442.1 | - | 5 | 436 | |||
JX502764.1 | - | 5 | 447 | |||
JX502765.1 | - | 5 | 436 |
Gene Symbol: SFTPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098668.3 | NP_001092138.1 | 5 | 460 | ||
NM_001320813.1 | NP_001307742.1 | 5 | 463 | |||
NM_001320814.1 | NP_001307743.1 | 4 | 529 | |||
XM_005270128.3 | XP_005270185.1 | 5 | 441 | |||
XM_005270132.3 | XP_005270189.1 | 4 | 428 | |||
XM_011540124.1 | XP_011538426.1 | 5 | 556 | |||
XM_011540125.1 | XP_011538427.1 | 5 | 499 | |||
XM_017016608.1 | XP_016872097.1 | 4 | 352 | |||
GenBank mRNA | AK225599.1 | - | 5 | 406 | ||
AK298041.1 | - | 5 | 313 | |||
AK298114.1 | - | 5 | 259 | |||
AK313179.1 | - | 5 | 409 | |||
AK316192.1 | - | 5 | 339 | |||
BC111571.1 | - | 5 | 385 | 40069329 | ||
BC139727.1 | - | 3 | 332 | 40024708 | ||
BC157866.1 | - | 5 | 399 | 9057085 | ||
BC157890.1 | - | 5 | 399 | 8862711 | ||
DC358710.1 | - | 4 | 193 | |||
HQ021421.1 | - | 5 | 409 | |||
HQ021422.2 | - | 5 | 409 | |||
HQ021423.2 | - | 5 | 409 | |||
HQ021424.1 | - | 5 | 409 | |||
HQ021425.2 | - | 5 | 409 | |||
HQ021426.1 | - | 5 | 406 | |||
HQ021427.1 | - | 5 | 406 | |||
HQ021428.2 | - | 5 | 406 | |||
HQ021429.2 | - | 5 | 406 | |||
HQ021430.1 | - | 5 | 406 | |||
HQ021431.2 | - | 5 | 406 | |||
HQ021432.1 | - | 5 | 409 | |||
K03475.1 | - | 5 | 397 |