Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313892.1 | NP_001300821.1 | 2 | 220 | ||
NM_001313893.1 | NP_001300822.1 | 2 | 980 | |||
NM_002128.5 | NP_002119.1 | 2 | 223 | |||
GenBank mRNA | AA430980.1 | - | 2 | 223 | ||
AB208998.1 | - | 2 | 107 | |||
AK057120.1 | - | 2 | 157 | |||
AK122825.1 | - | 2 | 982 | |||
AK291494.1 | - | 2 | 194 | |||
AK304506.1 | - | 2 | 194 | |||
AK310975.1 | - | 2 | 169 | |||
AV763680.1 | - | 2 | 220 | |||
AX795278.1 | - | 2 | 115 | |||
AY377859.1 | - | 1 | 39 | |||
BC003378.1 | - | 2 | 115 | 2901382 | ||
BC030981.1 | - | 2 | 134 | 4041682 | ||
BC066889.1 | - | 2 | 118 | 5268698 | ||
BC067732.1 | - | 2 | 138 | 4818524 | ||
BC141844.1 | - | 2 | 116 | 8860319 | ||
BT006940.1 | - | 1 | 39 | |||
BT020159.1 | - | 1 | 39 | |||
BX647267.1 | - | 1 | 603 | |||
CB051582.1 | - | 2 | 231 | |||
CR456863.1 | - | 1 | 39 | |||
CR749614.1 | - | 2 | 211 | |||
X12597.1 | - | 2 | 91 |
Gene Symbol: ACYP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017005408.1 | XP_016860897.1 | 2 | 383 |
Gene Symbol: CTCFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC137482.1 | - | 1 | 74 | 9021482 | |
BC137483.1 | - | 1 | 74 | 9021483 | ||
DQ778130.1 | - | 4 | 579 | |||
DQ778131.1 | - | 3 | 439 |