Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTCFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC137482.1 | - | 1 | 207 | 9021482 | |
BC137483.1 | - | 1 | 207 | 9021483 | ||
DQ778130.1 | - | 4 | 712 | |||
DQ778131.1 | - | 3 | 572 |
Gene Symbol: HMGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313892.1 | NP_001300821.1 | 3 | 353 | ||
NM_001313893.1 | NP_001300822.1 | 3 | 1113 | |||
NM_002128.5 | NP_002119.1 | 3 | 356 | |||
GenBank mRNA | AB208998.1 | - | 3 | 240 | ||
AK122825.1 | - | 3 | 1115 | |||
AK291494.1 | - | 3 | 327 | |||
AK304506.1 | - | 3 | 327 | |||
AK310975.1 | - | 3 | 302 | |||
AV763680.1 | - | 3 | 353 | |||
AX795278.1 | - | 3 | 248 | |||
AY377859.1 | - | 2 | 172 | |||
BC003378.1 | - | 3 | 248 | 2901382 | ||
BC030981.1 | - | 3 | 267 | 4041682 | ||
BC066889.1 | - | 3 | 251 | 5268698 | ||
BC067732.1 | - | 3 | 271 | 4818524 | ||
BC141844.1 | - | 3 | 249 | 8860319 | ||
BT006940.1 | - | 2 | 172 | |||
BT020159.1 | - | 2 | 172 | |||
BX647267.1 | - | 2 | 736 | |||
CB051582.1 | - | 3 | 364 | |||
CR456863.1 | - | 2 | 172 | |||
CR749614.1 | - | 3 | 344 | |||
D63874.1 | - | 3 | 248 | |||
X12597.1 | - | 3 | 224 |
Gene Symbol: SP100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003113.3 | NP_003104.2 | 25 | 2335 | ||
GenBank mRNA | AF056322.1 | - | 25 | 2225 | ||
L79988.1 | - | 10 | 766 |