Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014.4 | NP_001005.1 | 6 | 603 | ||
NM_001203245.2 | NP_001190174.1 | 6 | 755 | |||
NM_001204091.1 | NP_001191020.1 | 6 | 607 | |||
GenBank mRNA | AB209165.1 | - | 1 | 558 | ||
BC001032.2 | - | 6 | 528 | 3139393 | ||
BC001955.1 | - | 6 | 582 | 2905318 | ||
BC005012.1 | - | 6 | 582 | 3637658 | ||
BC070235.1 | - | 6 | 526 | 6453788 | ||
BC071946.1 | - | 6 | 538 | 6305105 | ||
BC073799.1 | - | 6 | 615 | 5517455 | ||
BE397113.1 | - | 6 | 553 | |||
BQ072109.1 | - | 1 | 723 | |||
BU956627.1 | - | 6 | 531 | |||
U14972.1 | - | 1 | 525 |
Gene Symbol: RPS10P19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK025353.1 | - | 1 | 3137 |