Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313892.1 | NP_001300821.1 | 3 | 402 | ||
NM_001313893.1 | NP_001300822.1 | 3 | 1162 | |||
NM_002128.5 | NP_002119.1 | 3 | 405 | |||
GenBank mRNA | AB208998.1 | - | 3 | 289 | ||
AK057120.1 | - | 3 | 268 | |||
AK122825.1 | - | 3 | 1164 | |||
AK291494.1 | - | 3 | 376 | |||
AK304506.1 | - | 3 | 376 | |||
AK310975.1 | - | 3 | 351 | |||
AV763680.1 | - | 3 | 402 | |||
AX795278.1 | - | 3 | 297 | |||
AY377859.1 | - | 2 | 221 | |||
BC003378.1 | - | 3 | 297 | 2901382 | ||
BC030981.1 | - | 3 | 316 | 4041682 | ||
BC066889.1 | - | 3 | 300 | 5268698 | ||
BC067732.1 | - | 3 | 320 | 4818524 | ||
BC141844.1 | - | 3 | 298 | 8860319 | ||
BT006940.1 | - | 2 | 221 | |||
BT020159.1 | - | 2 | 221 | |||
BX647267.1 | - | 2 | 785 | |||
CB051582.1 | - | 3 | 413 | |||
CR456863.1 | - | 2 | 221 | |||
CR749614.1 | - | 3 | 393 | |||
D63874.1 | - | 3 | 297 | |||
X12597.1 | - | 3 | 273 |
Gene Symbol: SP100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003113.3 | NP_003104.2 | 25 | 2384 | ||
GenBank mRNA | AF056322.1 | - | 25 | 2274 | ||
L79988.1 | - | 10 | 815 |