Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313892.1 | NP_001300821.1 | 5 | 712 | ||
NM_001313893.1 | NP_001300822.1 | 5 | 1472 | |||
NM_002128.5 | NP_002119.1 | 5 | 715 | |||
GenBank mRNA | AB208998.1 | - | 4 | 1604 | ||
AK057120.1 | - | 5 | 578 | |||
AK122825.1 | - | 5 | 1474 | |||
AK291494.1 | - | 5 | 686 | |||
AK310975.1 | - | 5 | 533 | |||
AX795278.1 | - | 5 | 607 | |||
AY377859.1 | - | 4 | 531 | |||
BC003378.1 | - | 5 | 607 | 2901382 | ||
BC030981.1 | - | 5 | 626 | 4041682 | ||
BC066889.1 | - | 5 | 610 | 5268698 | ||
BC067732.1 | - | 5 | 630 | 4818524 | ||
BC141844.1 | - | 5 | 608 | 8860319 | ||
BT006940.1 | - | 4 | 531 | |||
BT020159.1 | - | 4 | 531 | |||
BX647267.1 | - | 5 | 1145 | |||
CR456863.1 | - | 4 | 531 | |||
CR749614.1 | - | 5 | 703 | |||
D63874.1 | - | 5 | 607 | |||
X12597.1 | - | 5 | 583 |
Gene Symbol: SP100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003113.3 | NP_003104.2 | 25 | 2694 | ||
GenBank mRNA | AF056322.1 | - | 25 | 2584 | ||
L79988.1 | - | 10 | 1125 |