Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000971.3 | NP_000962.2 | 1 | 507 | ||
XM_006716463.2 | XP_006716526.1 | 5 | 1100 | |||
GenBank mRNA | AK225246.1 | - | 1 | 506 | ||
AK225254.1 | - | 1 | 505 | |||
AK291119.1 | - | 5 | 505 | |||
AK313365.1 | - | 5 | 505 | |||
BC006095.1 | - | 1 | 495 | 2821364 | ||
BC008850.2 | - | 1 | 532 | 4138113 | ||
BC009599.1 | - | 1 | 507 | 4134557 | ||
BC071671.1 | - | 1 | 488 | 4392472 | ||
BC071894.1 | - | 1 | 492 | 2906310 | ||
BC071895.1 | - | 1 | 490 | 6568654 | ||
BC087837.1 | - | 1 | 491 | 6170371 | ||
CR456773.1 | - | 1 | 485 | |||
L16558.1 | - | 1 | 485 | |||
X52967.1 | - | 5 | 506 | |||
X57959.1 | - | 1 | 495 |