Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008801.1 | NP_001008801.1 | 4 | 963 | ||
NM_001277120.1 | NP_001264049.1 | 5 | 1081 | |||
NR_102299.1 | 6 | 1336 | ||||
NR_102300.1 | 6 | 1317 | ||||
NR_102301.1 | 5 | 1218 | ||||
XM_017027443.1 | XP_016882932.1 | 5 | 1178 | |||
XM_017027444.1 | XP_016882933.1 | 6 | 1296 | |||
GenBank mRNA | AK025501.1 | - | 1 | 189 | ||
AK289841.1 | - | 5 | 1072 | |||
AK292449.1 | - | 4 | 932 | |||
AY442330.1 | - | 4 | 964 | |||
AY442331.1 | - | 5 | 1082 | |||
CR749381.1 | - | 1 | 57 | |||
CR936599.1 | - | 4 | 929 | |||
CR936695.1 | - | 4 | 933 |
Gene Symbol: LOC107983997 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011527096.2 | XP_011525398.1 | 4 | 1468 |
Gene Symbol: ZNF137P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023311.1 | 1 | 237 | |||
GenBank mRNA | BC110455.1 | - | 1 | 237 | ||
BC110456.1 | - | 1 | 237 | |||
U09414.1 | - | 1 | 271 |
Gene Symbol: ZNF160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102603.1 | NP_001096073.1 | 7 | 1438 | ||
NM_001322128.1 | NP_001309057.1 | 7 | 1446 | |||
NM_001322129.1 | NP_001309058.1 | 6 | 1541 | |||
NM_001322130.1 | NP_001309059.1 | 7 | 1670 | |||
NM_001322131.1 | NP_001309060.1 | 6 | 1545 | |||
NM_001322132.1 | NP_001309061.1 | 7 | 1434 | |||
NM_001322133.1 | NP_001309062.1 | 7 | 1458 | |||
NM_001322134.1 | NP_001309063.1 | 7 | 1462 | |||
NM_001322135.1 | NP_001309064.1 | 6 | 1317 | |||
NM_001322136.1 | NP_001309065.1 | 5 | 1174 | |||
NM_001322137.1 | NP_001309066.1 | 8 | 1494 | |||
NM_001322138.1 | NP_001309067.1 | 7 | 1377 | |||
NM_001322139.1 | NP_001309068.1 | 9 | 1591 | |||
NM_033288.3 | NP_150630.2 | 6 | 1321 | |||
NM_198893.2 | NP_942596.1 | 7 | 1406 | |||
XM_017027445.1 | XP_016882934.1 | 4 | 1670 | |||
XM_017027446.1 | XP_016882935.1 | 6 | 1631 | |||
GenBank mRNA | AB046831.1 | - | 3 | 1791 | ||
AF277623.1 | - | 6 | 1329 | |||
AK024442.1 | - | 1 | 685 | |||
BC094880.1 | - | 7 | 1323 | 3066532 | ||
CR749613.1 | - | 1 | 136 | |||
X77744.1 | - | 1 | 248 | |||
X78928.1 | - | 1, 2 | 398 |
Gene Symbol: ZNF28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006969.3 | NP_008900.3 | 4 | 969 | ||
NR_036599.1 | 5 | 1087 | ||||
NR_036600.1 | 3 | 842 | ||||
XM_006723354.3 | XP_006723417.1 | 4 | 944 | |||
XM_011527262.2 | XP_011525564.1 | 4 | 999 | |||
GenBank mRNA | AK091553.1 | - | 1 | 53 | ||
AK092385.1 | - | 1 | 28 | |||
AK131368.1 | - | 2 | 1567 | |||
AK131574.1 | - | 1 | 354 | |||
AK293074.1 | - | 3 | 842 | |||
AK304704.1 | - | 4 | 933 | |||
BC140720.1 | - | 3 | 870 | 9021588 | ||
BX640767.1 | - | 1 | 248 | |||
CR933598.1 | - | 4 | 932 |
Gene Symbol: ZNF347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172674.1 | NP_001166145.1 | 5 | 1548 | ||
NM_001172675.1 | NP_001166146.1 | 5 | 1228 | |||
NM_032584.2 | NP_115973.2 | 5 | 1225 | |||
XM_005259335.4 | XP_005259392.1 | 5 | 1547 | |||
XM_005259336.4 | XP_005259393.1 | 5 | 1337 | |||
XM_017027384.1 | XP_016882873.1 | 5 | 1567 | |||
XM_017027385.1 | XP_016882874.1 | 5 | 1251 | |||
XM_017027386.1 | XP_016882875.1 | 4 | 1173 | |||
GenBank mRNA | AK095593.1 | - | 1 | 786 | ||
AK096623.1 | - | 5 | 1550 | |||
AK289994.1 | - | 5 | 1212 | |||
AK314603.1 | - | 5 | 1209 | |||
AL713691.1 | - | 1 | 207 | |||
AY029765.1 | - | 5 | 1187 | |||
BC136255.1 | - | 4 | 1177 | 9020242 | ||
BC143412.1 | - | 4 | 1180 |
Gene Symbol: ZNF354C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014594.2 | NP_055409.1 | 5 | 1121 | ||
XM_017009409.1 | XP_016864898.1 | 5 | 1224 | |||
GenBank mRNA | AF395540.1 | - | 3 | 788 | ||
AF525463.1 | - | 5 | 930 | |||
AL137483.1 | - | 1 | 111 | |||
BC063312.1 | - | 5 | 1121 | 30341631 |
Gene Symbol: ZNF525 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003699.1 | 4 | 897 | |||
GenBank mRNA | AB075859.1 | - | 1 | 1351 | ||
AK098804.1 | - | 1 | 1476 |
Gene Symbol: ZNF578 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099694.1 | NP_001093164.1 | 6 | 1104 | ||
XM_017026302.1 | XP_016881791.1 | 6 | 1124 | |||
GenBank mRNA | AK055946.1 | - | 1 | 282 | ||
AK095562.1 | - | 1 | 219 | |||
AK299106.1 | - | 6 | 1104 | |||
BC103904.2 | - | 1 | 272 | |||
BC115704.1 | - | 1 | 272 |
Gene Symbol: ZNF600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321866.1 | NP_001308795.1 | 6 | 1699 | ||
NM_001321867.1 | NP_001308796.1 | 4 | 1076 | |||
NM_198457.3 | NP_940859.3 | 3 | 949 | |||
XM_017026390.1 | XP_016881879.1 | 7 | 1626 | |||
XM_017026391.1 | XP_016881880.1 | 6 | 1602 | |||
XM_017026392.1 | XP_016881881.1 | 6 | 1400 | |||
GenBank mRNA | AK075230.1 | - | 1 | 236 | ||
AK131231.1 | - | 3 | 939 | |||
BX640933.1 | - | 3 | 921 |
Gene Symbol: ZNF611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161499.1 | NP_001154971.1 | 6 | 1972 | ||
NM_001161500.1 | NP_001154972.1 | 5 | 1872 | |||
NM_001161501.1 | NP_001154973.1 | 5 | 1835 | |||
NM_030972.3 | NP_112234.3 | 7 | 2014 | |||
GenBank mRNA | AK021787.1 | - | 1 | 93 | ||
AK091389.1 | - | 2 | 1552 | |||
AK097434.1 | - | 7 | 2014 | |||
AL834123.1 | - | 1 | 1376 | |||
BC028590.1 | - | 9 | 2256 | |||
BC136343.1 | - | 5 | 1920 | 9020330 | ||
BC143996.1 | - | 4 | 1769 |
Gene Symbol: ZNF665 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024733.3 | NP_079009.3 | 4 | 885 | ||
XM_005259266.4 | XP_005259323.1 | 4 | 919 | |||
XM_011527325.2 | XP_011525627.1 | 4 | 990 | |||
XM_011527327.2 | XP_011525629.1 | 5 | 1294 | |||
GenBank mRNA | AK291403.1 | - | 4 | 875 | ||
AL832507.1 | - | 4 | 870 |
Gene Symbol: ZNF701 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172655.1 | NP_001166126.1 | 5 | 1316 | ||
NM_018260.2 | NP_060730.2 | 4 | 1022 | |||
XM_011527092.1 | XP_011525394.1 | 4 | 1096 | |||
GenBank mRNA | AK001753.1 | - | 4 | 1020 | ||
BC080612.1 | - | 3 | 978 | 5816672 | ||
BC131719.1 | - | 4 | 996 | 40123413 | ||
BC136425.1 | - | 4 | 1126 | 9020414 | ||
BC136426.1 | - | 4 | 994 | 9020415 |
Gene Symbol: ZNF702P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003578.1 | 4 | 1051 | |||
GenBank mRNA | AK023047.1 | - | 1 | 264 | ||
AK092593.1 | - | 4 | 1051 | |||
AL832505.1 | - | 4 | 1048 | |||
BC032590.1 | - | 1 | 430 |
Gene Symbol: ZNF761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008401.3 | NP_001008401.3 | 6 | 1519 | ||
NM_001289951.1 | NP_001276880.1 | 5 | 1300 | |||
NM_001289952.1 | NP_001276881.1 | 5 | 1368 | |||
GenBank mRNA | AB107355.2 | - | 5 | 1369 | ||
AK092652.1 | - | 1 | 777 | |||
AK131281.1 | - | 2 | 989 |
Gene Symbol: ZNF765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040185.1 | NP_001035275.1 | 4 | 935 | ||
GenBank mRNA | AK293890.1 | - | 6 | 1149 | ||
AK316300.1 | - | 4 | 929 | |||
BC140724.1 | - | 4 | 909 | 9021592 | ||
BX647981.1 | - | 4 | 927 |
Gene Symbol: ZNF808 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039886.3 | NP_001034975.2 | 5 | 1205 | ||
NM_001321424.1 | NP_001308353.1 | 5 | 1350 | |||
NM_001321425.1 | NP_001308354.1 | 4 | 1103 | |||
XM_005258909.3 | XP_005258966.1 | 4 | 1066 | |||
GenBank mRNA | CR749856.1 | - | 3 | 981 |
Gene Symbol: ZNF813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004301.3 | NP_001004301.2 | 4 | 1612 | ||
GenBank mRNA | AK091460.1 | - | 1 | 760 | ||
AK131422.1 | - | 1 | 995 |
Gene Symbol: ZNF816 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031665.2 | NP_001026835.1 | 5 | 1289 | ||
NM_001202456.1 | NP_001189385.1 | 4 | 1148 | |||
NM_001202457.1 | NP_001189386.1 | 4 | 1138 | |||
GenBank mRNA | AK291990.1 | - | 4 | 1052 | ||
AL713804.1 | - | 1 | 557 | |||
BC105739.1 | - | 4 | 995 | 40026775 | ||
BC105740.1 | - | 4 | 998 | 40026776 | ||
BC105930.1 | - | 4 | 995 | 40026773 |
Gene Symbol: ZNF83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054659.1 | - | 1 | 55 |
Gene Symbol: ZNF845 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321522.1 | NP_001308451.1 | 5 | 1450 | ||
NM_001321523.1 | NP_001308452.1 | 5 | 1389 | |||
NM_001321524.1 | NP_001308453.1 | 5 | 1397 | |||
NM_138374.2 | NP_612383.1 | 4 | 1295 | |||
GenBank mRNA | AK096653.1 | - | 1 | 642 | ||
AK295079.1 | - | 5 | 1369 | |||
BC007307.1 | - | 1 | 151 |
Gene Symbol: ZNF860 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137674.2 | NP_001131146.2 | 2 | 1406 | ||
XM_017006299.1 | XP_016861788.1 | 2 | 2489 | |||
GenBank mRNA | AK294003.1 | - | 2 | 1406 |
Gene Symbol: ZNF888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310127.1 | NP_001297056.1 | 3 | 1035 | ||
XM_005259451.4 | XP_005259508.1 | 5 | 1267 | |||
XM_017026798.1 | XP_016882287.1 | 5 | 1428 | |||
XM_017026799.1 | XP_016882288.1 | 5 | 1313 | |||
XM_017026800.1 | XP_016882289.1 | 3 | 1044 |