Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002995.2 | NP_002986.1 | 3 | 628 | ||
XM_011509865.2 | XP_011508167.1 | 4 | 2853 | |||
GenBank mRNA | BC070309.1 | - | 3 | 496 | 30416260 | |
D43768.1 | - | 3 | 483 | |||
U23772.1 | - | 3 | 477 | |||
X86474.1 | - | 3 | 487 |
Gene Symbol: XCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003175.3 | NP_003166.1 | 3 | 496 |