Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TECR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321170.1 | NP_001308099.1 | 14 | 1039 | ||
NM_138501.5 | NP_612510.1 | 13 | 965 | |||
NR_038103.1 | 13 | 997 | ||||
NR_038104.1 | 13 | 1030 | ||||
XM_006722945.1 | XP_006723008.1 | 14 | 1083 | |||
XM_011528442.2 | XP_011526744.1 | 13 | 913 | |||
GenBank mRNA | AF038958.1 | - | 13 | 911 | ||
AF038959.1 | - | 8 | 394 | |||
AF222742.1 | - | 13 | 886 | |||
AK001416.1 | - | 13 | 929 | |||
AK094087.1 | - | 14 | 1006 | |||
AK299134.1 | - | 14 | 1003 | |||
AK315412.1 | - | 13 | 929 | |||
BC000174.2 | - | 12 | 825 | |||
BC002425.1 | - | 13 | 897 | 3163592 | ||
BC005952.1 | - | 13 | 908 | 4249841 | ||
BC007801.2 | - | 13 | 915 | 4128702 | ||
BC013881.2 | - | 13 | 913 | 3928698 | ||
BT007179.1 | - | 12 | 835 | |||
CR456892.1 | - | 12 | 835 |