Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278697.1 | NP_001265626.1 | 2 | 192 | ||
NM_003147.5 | NP_003138.3 | 2 | 192 | |||
NM_175698.2 | NP_783629.1 | 2 | 192 | |||
GenBank mRNA | BC002818.2 | - | 2 | 126 | 3636268 | |
BC007343.2 | - | 2 | 126 | 3615133 | ||
BC016957.1 | - | 2 | 126 | 3916128 | ||
BC069313.1 | - | 1 | 56 | 7262129 | ||
BC071827.1 | - | 2 | 126 | 4641362 | ||
BC103863.2 | - | 2 | 194 | 40003337 | ||
BG104299.1 | - | 2 | 164 | |||
BI560469.1 | - | 2 | 199 | |||
JX014236.1 | - | 1 | 56 | |||
X86175.1 | - | 2 | 147 | |||
Z49105.1 | - | 2 | 154 | |||
Z49106.1 | - | 1 | 86 |
Gene Symbol: SSX2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164417.2 | NP_001157889.1 | 2 | 191 | ||
NM_001278701.1 | NP_001265630.1 | 2 | 191 | |||
NM_001278702.1 | NP_001265631.1 | 2 | 191 | |||
GenBank mRNA | BE391023.1 | - | 2 | 145 | ||
CB158955.1 | - | 2 | 108 |
Gene Symbol: SSX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173358.2 | NP_775494.1 | 2 | 215 | ||
XM_017029430.1 | XP_016884919.1 | 1 | 56 | |||
XM_017029431.1 | XP_016884920.1 | 1 | 56 |
Gene Symbol: SSX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073393.1 | 2 | 143 | |||
GenBank mRNA | DB029485.1 | - | 2 | 143 |