Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037290.2 | NP_001032367.2 | 12 | 2566 | ||
NM_001256397.1 | NP_001243326.1 | 12 | 2548 | |||
NM_001256398.1 | NP_001243327.1 | 10 | 2459 | |||
NM_001256400.1 | NP_001243329.1 | 11 | 2361 | |||
NM_031938.5 | NP_114144.4 | 12 | 2678 |