Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP1LC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022818.4 | NP_073729.1 | 4 | 543 | ||
GenBank mRNA | AF087871.1 | - | 4 | 402 | ||
AF183417.1 | - | 4 | 402 | |||
AF303888.1 | - | 4 | 348 | |||
AK025556.1 | - | 4 | 394 | |||
AK290956.1 | - | 4 | 425 | |||
AK310224.1 | - | 5 | 510 | |||
AL834298.1 | - | 4 | 312 | |||
BC018634.2 | - | 4 | 381 | 3623259 | ||
BC041874.1 | - | 4 | 938 | 5271912 | ||
BC045759.1 | - | 5 | 884 | 4828857 | ||
BC048009.1 | - | 4 | 427 | |||
BC067797.1 | - | 4 | 394 | 5276841 | ||
DA577457.1 | - | 4 | 423 | |||
JN663879.1 | - | 4 | 334 |
Gene Symbol: MAP1LC3B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085481.1 | NP_001078950.1 | 2 | 464 | ||
XM_011538652.2 | XP_011536954.1 | 2 | 637 |