Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001807.4 | NP_001798.2 | 8 | 990 | ||
GenBank mRNA | AF081673.1 | - | 7 | 905 | ||
AK299965.1 | - | 8 | 867 | |||
BC042510.1 | - | 8 | 975 | 5187959 | ||
M85201.1 | - | 8 | 982 | |||
S79774.1 | - | 8 | 984 | |||
X54457.1 | - | 8 | 1055 |
Gene Symbol: CELP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001275.2 | 2 | 158 | |||
GenBank mRNA | L14813.1 | - | 2 | 158 |