Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006908.4 | NP_008839.2 | 1, 2 | 280 | ||
NM_018890.3 | NP_061485.1 | 1, 2 | 280 | |||
GenBank mRNA | AB209410.1 | - | 1, 2 | 227 | ||
AB451238.1 | - | 1, 2 | 39 | |||
AB451363.1 | - | 1, 2 | 39 | |||
AF136373.1 | - | 1, 2 | 39 | |||
AF498964.1 | - | 1, 2 | 39 | |||
AJ132694.1 | - | 1, 2 | 39 | |||
AK054993.1 | - | 1, 2 | 255 | |||
AK291806.1 | - | 1, 2 | 236 | |||
AK291815.1 | - | 1, 2 | 226 | |||
AY279384.1 | - | 1, 2 | 197 | |||
BC004247.1 | - | 1, 2 | 177 | 3610745 | ||
BC050687.1 | - | 1, 2 | 227 | 6149377 | ||
BC107748.1 | - | 1, 2 | 242 | 6699945 | ||
BG437715.1 | - | 1, 2 | 279 | |||
BT007121.1 | - | 1, 2 | 39 | |||
M29870.1 | - | 1, 2 | 39 | |||
M31467.1 | - | 1, 2 | 39 |