Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198924.3 | NP_944606.2 | 5 | 1011 | ||
GenBank mRNA | AF498998.1 | - | 5 | 1011 | ||
AK126632.1 | - | 2 | 3536 | |||
BC073926.1 | - | 5 | 1078 |
Gene Symbol: TRIM74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317815.1 | NP_001304744.1 | 5 | 950 | ||
NM_198853.2 | NP_942150.1 | 5 | 1011 | |||
XM_011516187.2 | XP_011514489.1 | 5 | 919 | |||
XM_011516189.2 | XP_011514491.1 | 5 | 1232 | |||
GenBank mRNA | AF498999.1 | - | 5 | 1011 | ||
BC033871.1 | - | 5 | 947 | 5201777 |