Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016538.2 | NP_057622.1 | 7, 8 | 875 | ||
XM_006722284.2 | XP_006722347.1 | 5, 6 | 609 | |||
XM_011523580.1 | XP_011521882.1 | 6, 7 | 741 | |||
XM_011523581.2 | XP_011521883.1 | 6, 7 | 2478 | |||
GenBank mRNA | AF233395.1 | - | 7, 8 | 857 | ||
AK002027.1 | - | 5, 6 | 1655 | |||
AK094326.1 | - | 6, 7 | 940 | |||
AK131484.1 | - | 2, 3 | 1430 | |||
AK290265.1 | - | 7, 8 | 843 | |||
AL137626.1 | - | 4, 5 | 443 | |||
BC017305.2 | - | 7, 8 | 839 | 5087554 | ||
BC101791.1 | - | 7, 8 | 856 | 8069297 | ||
BC101793.1 | - | 7, 8 | 855 | 8069299 |