Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021965.3 | NP_068800.2 | 2 | 641 | ||
XM_011518782.2 | XP_011517084.1 | 2 | 931 | |||
XM_011518783.2 | XP_011517085.1 | 2 | 931 | |||
XM_017014808.1 | XP_016870297.1 | 2 | 533 | |||
GenBank mRNA | AK297108.1 | - | 2 | 463 | ||
BC033073.1 | - | 2 | 641 | 4123706 |
Gene Symbol: PGM5P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002836.2 | 2 | 589 | |||
GenBank mRNA | BC025351.1 | - | 2 | 581 |