Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA8F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033351.1 | 16 | 1930 | |||
GenBank mRNA | AK057241.1 | - | 14 | 1408 | ||
AL833871.1 | - | 5 | 410 | |||
BC144672.1 | - | 13 | 1535 | |||
BC150302.1 | - | 16 | 1904 | 8860399 | ||
DQ309038.1 | - | 14 | 1378 |
Gene Symbol: GOLGA8CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027411.2 | 16 | 1942 |
Gene Symbol: GOLGA8DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027407.1 | 16 | 1930 | |||
GenBank mRNA | AK292534.1 | - | 16 | 1813 | ||
BC110417.1 | - | 16 | 1901 | 5171794 |
Gene Symbol: GOLGA8EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033350.1 | 16 | 1930 | |||
GenBank mRNA | BC063309.1 | - | 17 | 1904 | ||
DQ309037.1 | - | 16 | 1904 |
Gene Symbol: GOLGA8G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033353.1 | 16 | 1942 | |||
GenBank mRNA | BC066774.1 | - | 16 | 1916 | ||
BC099913.1 | - | 15 | 2757 | 4838463 | ||
BC125193.1 | - | 15 | 1722 | 40122746 | ||
BC125194.1 | - | 15 | 1722 | 40122749 |