Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMPR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004329.2 | NP_004320.2 | 10 | 1479 | ||
XM_011540103.2 | XP_011538405.1 | 10 | 1376 | |||
XM_011540104.2 | XP_011538406.1 | 11 | 1543 | |||
GenBank mRNA | AK291764.1 | - | 10 | 1348 | ||
BC028383.1 | - | 10 | 1473 | 4824378 | ||
Z22535.1 | - | 10 | 1240 |